3-(2-Methoxyphenyl)propionic acid

Supplier: Matrix Scientific

037484-5G 037484-25G
101843-504EA 47.69 USD
101843-504 101843-506
3-(2-Methoxyphenyl)propionic acid
3-(2-Methoxyphenyl)propionic acid

Hazard code(s): Irritant

Formula: C₁₀H₁₂O₃
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 92 °C
MDL Number: MFCD00002772
CAS Number: 6342-77-4
EINECS: 228-738-0

Order Now

Specification Test Results

Melting point 90-92°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR