2-(2-Aminoethyl)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Pyridylethylamine, 2-(2-Pyridyl)ethylamine, 2-Pyridin-2-ylethanamine

037478-5G 037478-25G
101843-488EA 21.54 USD
101843-488 101843-490
2-(2-Aminoethyl)pyridine ≥95%
2-(2-Aminoethyl)pyridine

Hazard code(s): Corrosive, Air Sensitive

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 208 °C
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006367
CAS Number: 2706-56-1
EINECS: 220-295-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR