2-Aminoisobutyric acid

Supplier: Matrix Scientific

Synonyms: α-Aminoisobutyric acid, 2-Amino-2-methylpropionic acid, 2-Metilalanina, H-Aib-OH, 2-Methylalanine, 2-Amino-2-methylpropanoic acid

037475-100G 037475-250G
101843-476EA 61.54 USD
101843-476 101843-478
2-Aminoisobutyric acid
2-Aminoisobutyric acid

Hazard code(s): Irritant

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: >300 °C
Density: 1.278 g/cm³ (20 °C)
MDL Number: MFCD00008049
CAS Number: 62-57-7
EINECS: 200-544-0
Merck Index: 13,00444

Order Now

Specification Test Results

Melting point >300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR