1-Isoquinolinecarboxylic acid

Supplier: Matrix Scientific

Synonyms: Isoquinoline-1-carboxylic acid

037459-5G 037459-25G
101843-436EA 15.38 USD
101843-436 101843-438
1-Isoquinolinecarboxylic acid
1-Isoquinolinecarboxylic acid

Hazard code(s): Irritant

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 161 °C
Storage Temperature: Ambient
MDL Number: MFCD00006901
CAS Number: 486-73-7
EINECS: 207-639-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR