4-Cyanobenzaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Formylbenzonitrile

037458-25G 037458-100G
101843-432EA 29.23 USD
101843-432 101843-434
4-Cyanobenzaldehyde ≥97%
4-Cyanobenzaldehyde

Hazard code(s): Irritant, Air Sensitive

Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00003376
CAS Number: 105-07-7
EINECS: 203-267-3
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR