1-(4-Fluorophenyl)piperazine ≥95%
Supplier: Matrix Scientific
037455-5G
037455-25G
101843-424EA
21.54
USD
101843-424
101843-426
1-(4-Fluorophenyl)piperazine ≥95%
1-(4-Fluorophenyl)piperazine
Hazard code(s): Irritant
Formula:
C₁₀H₁₃FN₂ MW: 180.23 g/mol Boiling Pt: 363 °C (1013 hPa) Melting Pt: 30…33 °C |
MDL Number:
MFCD00005960 CAS Number: 2252-63-3 EINECS: 218-846-6 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...