1-(3-Methoxyphenyl)piperazine for synthesis
Supplier: Matrix Scientific
037439-5G
037439-25G
101843-376EA
41.54
USD
101843-376
101843-378
1-(3-Methoxyphenyl)piperazine for synthesis
1-(3-Methoxyphenyl)piperazine
Formula:
C₁₁H₁₆N₂O Boiling Pt: 130 °C (0.5 mmHg) Density: 1.082 g/cm³ (20 °C) |
MDL Number:
MFCD00040733 CAS Number: 16015-71-7 EINECS: 240-154-8 UN: 3267 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...