Methyl-5-bromo-2-furoate
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2-furoic acid methyll ester, 5-Bromo-2-furancarboxylic acid methyll ester, Methyl 5-Bromo-2-furancarboxylate, Methyl 5-Bromo-2-furoate, Methyl 5-bromofuran-2-carboxylate
037169-5G
037169-25G
101842-818EA
41.54
USD
101842-818
101842-820
Methyl-5-bromo-2-furoate
Methyl-5-bromo-2-furoate
Formula:
C₆H₅BrO₃ MW: 205.01 g/mol |
MDL Number:
MFCD00092313 CAS Number: 2527-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...