3-Phenyl-1H-pyrazole ≥95%

Supplier: Matrix Scientific

037139-5G 037139-25G
101842-760EA 38.46 USD
101842-760 102551-204
3-Phenyl-1H-pyrazole ≥95%
3-Phenyl-1H-pyrazole

Hazard code(s): Irritant

Formula: C₉H₈N₂
Melting Pt: 75…82 °C
MDL Number: MFCD00159654
CAS Number: 2458-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR