2,4-Dimethoxybenzylamine ≥95%

Supplier: Matrix Scientific

037102-25G 037102-100G
101842-680EA 56.92 USD
101842-680 102551-196
2,4-Dimethoxybenzylamine ≥95%
2,4-Dimethoxybenzylamine

Hazard code(s): Corrosive, Irritant

Formula: C₉H₁₃NO₂
Boiling Pt: 140 °C (1mm)
Density: 1.113 g/cm³
MDL Number: MFCD00052393
CAS Number: 20781-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR