1-Methyl-4-piperidinemethanol
Supplier: Matrix Scientific
037045-5G
037045-25G
101842-560EA
21.54
USD
101842-560
101842-562
1-Methyl-4-piperidinemethanol
1-Methyl-4-piperidinemethanol
Hazard code(s): Irritant
Formula:
C₇H₁₅NO Boiling Pt: 79…80 °C |
MDL Number:
MFCD03411903 CAS Number: 20691-89-8 |
Specification Test Results
Boiling point | 79-80°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...