3-Nitro-1H-pyrazole ≥95%

Supplier: Matrix Scientific

036455-5G 036455-25G
101841-404EA 36.92 USD
101841-404 102551-188
3-Nitro-1H-pyrazole ≥95%
3-Nitro-1H-pyrazole

Hazard code(s): Irritant

Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
MDL Number: MFCD00159621
CAS Number: 26621-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR