1-Methyl-1H-pyrazole ≥98%
Supplier: Matrix Scientific
|
Danger
|
036425-25G
036425-100G
101841-334EA
44.62
USD
101841-334
101841-336
1-Methyl-1H-pyrazole ≥98%
1-Methyl-1H-pyrazole
Hazard code(s): Irritant
Formula:
C₄H₆N₂ Boiling Pt: 127 °C (1013 hPa) Density: 0.993 g/cm³ (20 °C) |
MDL Number:
MFCD00144943 CAS Number: 930-36-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...