Rhodanine

Supplier: Matrix Scientific

Synonyms: 2-Thioxo-4-thiazolidinone, 2-Thioxo-1,3-thiazolidin-4-one, 2-Thioxothiazolidin-4-one

035973-25G 035973-100G
101840-436EA 29.23 USD
101840-436 101840-438
Rhodanine
Rhodanine

Hazard code(s): Irritant

Formula: C₃H₃NOS₂
MW: 133.19 g/mol
Melting Pt: 166…171 °C
MDL Number: MFCD00005488
CAS Number: 141-84-4
EINECS: 205-505-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR