5-Fluoro-2-hydroxypyridine ≥95% for synthesis

Supplier: Matrix Scientific

035690-1G 035690-5G
101839-688EA 29.23 USD
101839-688 101839-690
5-Fluoro-2-hydroxypyridine ≥95% for synthesis
5-Fluoro-2-hydroxypyridine

Hazard code(s): Irritant

Formula: C₅H₄FNO
MDL Number: MFCD03092918
CAS Number: 51173-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR