4-(Indol-3-yl)butyric acid ≥98%

Supplier: Matrix Scientific

035689-25G 035689-100G
101839-684EA 21.54 USD
101839-684 101839-686
4-(Indol-3-yl)butyric acid ≥98%
4-(Indol-3-yl)butyric acid

Hazard code(s): Irritant, Moisture Sensitive

Formula: C₁₂H₁₃NO₂
Melting Pt: 121…125 °C
Density: 1.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005664
CAS Number: 133-32-4
EINECS: 205-101-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,04987

Order Now

Specification Test Results

Melting point 121-125°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR