4-Hydroxyphenylboronic acid pinacol ester ≥97%
Supplier: Matrix Scientific
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 4-Hydroxybenzeneboronic acid pinacol ester
035607-5G
035607-25G
101839-564EA
53.85
USD
101839-564
101839-566
4-Hydroxyphenylboronic acid pinacol ester ≥97%
4-Hydroxyphenylboronic acid pinacol ester
Hazard code(s): Irritant, Store in Freezer
Formula:
C₁₂H₁₇BO₃ MW: 220.08 g/mol Melting Pt: 113…117 °C |
MDL Number:
MFCD02093756 CAS Number: 269409-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...