4-Chloro-3-(trifluoromethyl)phenylboronic acid
Supplier: Matrix Scientific
Synonyms:
Dihydroxy(4-chloro-3-(trifluoromethyl)phenyl)borane, 4-Chloro-3-(trifluoromethyl)benzeneboronic acid, (4-Chloro-3-(trifluoromethyl)phenyl)metaboric acid
035574-5G
035574-25G
101839-528EA
56.92
USD
101839-528
101839-530
4-Chloro-3-(trifluoromethyl)phenylboronic acid
4-Chloro-3-(trifluoromethyl)phenylboronic acid
Formula:
C₇H₅BClF₃O₂ MW: 224.37 g/mol |
MDL Number:
MFCD03094999 CAS Number: 176976-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...