4-Chloro-2-fluorophenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-fluorobenzeneboronic acid, (4-Chloro-2-fluorophenyl)metaboric acid, Dihydroxy(4-chloro-2-fluorophenyl)borane

035571-5G 035571-25G
101839-524EA 35.38 USD
101839-524 101839-526
4-Chloro-2-fluorophenylboronic acid ≥95%
4-Chloro-2-fluorophenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₅BClFO₂
MW: 174.37 g/mol
Melting Pt: 248…250 °C
MDL Number: MFCD02684293
CAS Number: 160591-91-3

Order Now

Specification Test Results

Melting point 248-250°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR