3-Hydroxyphenylboronic acid pinacol ester

Supplier: Matrix Scientific

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

035535-1G 035535-5G
101839-484EA 106.15 USD
101839-484 101839-486
3-Hydroxyphenylboronic acid pinacol ester
3-Hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₇BO₃
MW: 220.08 g/mol
MDL Number: MFCD02093755
CAS Number: 214360-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR