3-Ethoxyphenylboronic acid

Supplier: Matrix Scientific

035521-5G 035521-25G
101839-480EA 60 USD
101839-480 101839-482
3-Ethoxyphenylboronic acid
3-Ethoxyphenylboronic acid
Formula: C₈H₁₁BO₃
MDL Number: MFCD00274219
CAS Number: 90555-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR