3,5-Dimethoxyphenylboronic acid

Supplier: Matrix Scientific

035484-5G 035484-25G
101839-460EA 46.15 USD
101839-460 101839-462
3,5-Dimethoxyphenylboronic acid
3,5-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MDL Number: MFCD03095127
CAS Number: 192182-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR