2-Fluoro-5-nitrophenylboronic acid for synthesis

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-5-nitrobenzeneboronic acid

035460-1G 035460-5G
101839-434EA 96.92 USD
101839-434 101839-436
2-Fluoro-5-nitrophenylboronic acid for synthesis
2-Fluoro-5-nitrophenylboronic acid
Formula: C₆H₅BFNO₄
MDL Number: MFCD03092929
CAS Number: 819849-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR