2-Chloro-4-(trifluoromethyl)phenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (2-Chloro-4-(trifluoromethyl)phenyl)metaboric acid, Dihydroxy(2-chloro-4-(trifluoromethyl)phenyl)borane, 2-Chloro-4-(trifluoromethyl)benzeneboronic acid

035386-1G 035386-5G 035386-25G
101839-384EA 33.85 USD
101839-384 101839-386 101839-388
2-Chloro-4-(trifluoromethyl)phenylboronic acid ≥95%
2-Chloro-4-(trifluoromethyl)phenylboronic acid

Hazard code(s): Irritant

Formula: C₇H₅BClF₃O₂
MW: 224.37 g/mol
MDL Number: MFCD02684337
CAS Number: 254993-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR