2,6-Difluoro-4-hydroxyphenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (2,6-Difluoro-4-hydroxyphenyl)boronic acid

035320-1G 035320-5G
101839-372EA 138.46 USD
101839-372 101839-374
2,6-Difluoro-4-hydroxyphenylboronic acid ≥95%
2,6-Difluoro-4-hydroxyphenylboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₆H₅BF₂O₃
MW: 173.91 g/mol
MDL Number: MFCD09878344
CAS Number: 957065-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR