2,5-Dichlorophenylboronic acid

Supplier: Matrix Scientific

035309-5G 035309-25G
101839-366EA 153.85 USD
101839-366 101839-368
2,5-Dichlorophenylboronic acid
2,5-Dichlorophenylboronic acid

Hazard code(s): Irritant, REFRIGERATE

Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Melting Pt: ∼210…216 °C
MDL Number: MFCD01863182
CAS Number: 135145-90-3

Order Now

Specification Test Results

Melting point ca 210-216

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR