2,4-Dimethylphenylboronic acid

Supplier: Matrix Scientific
Warning

035306-5G 035306-25G
101839-362EA 32.31 USD
101839-362 101839-364
2,4-Dimethylphenylboronic acid
2,4-Dimethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
MDL Number: MFCD02683101
CAS Number: 55499-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR