2,4-Dichlorophenylboronic acid

Supplier: Matrix Scientific

035304-5G 035304-25G
101839-356EA 21.54 USD
101839-356 101839-358
2,4-Dichlorophenylboronic acid
2,4-Dichlorophenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₅BCl₂O₂
Melting Pt: 242…245 °C
MDL Number: MFCD00013930
CAS Number: 68716-47-2

Order Now

Specification Test Results

Melting point 242-245 de


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR