1-Methyl-1H-pyrazole-4-boronic acid pinacol ester for synthesis

Supplier: Matrix Scientific

Synonyms: 1-Methylpyrazole-4-boronic acid pinacol ester, 1-Methyl-4-pyrazole boronic acid pinacol ester, 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

035281-5G 035281-25G
101839-352EA 29.23 USD
101839-352 101839-354
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester for synthesis
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester

Hazard code(s): Irritant

Formula: C₁₀H₁₇BN₂O₂
Melting Pt: 64…68 °C
MDL Number: MFCD03789259
CAS Number: 761446-44-0

Order Now

Specification Test Results

Melting point 64-68°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR