1-Butylboronic acid

Supplier: Matrix Scientific

Synonyms: Butylboronic acid, n-Butylboronic acid

035280-25G 035280-100G
101839-348EA 130.77 USD
101839-348 101839-350
1-Butylboronic acid
1-Butylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₄H₁₁BO₂
MW: 101.94 g/mol
Melting Pt: 94…96 °C
MDL Number: MFCD00002106
CAS Number: 4426-47-5
EINECS: 224-607-7

Order Now

Specification Test Results

Melting point 94-96 ºC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR