4,5-Diphenyl-1,2,3-thiadiazole ≥97%
Supplier: Matrix Scientific
035277-1G
035277-5G
101839-336EA
80
USD
101839-336
101839-338
4,5-Diphenyl-1,2,3-thiadiazole ≥97%
4,5-Diphenyl-1,2,3-thiadiazole
Hazard code(s): Irritant
Formula:
C₁₄H₁₀N₂S Melting Pt: 92…94 °C |
MDL Number:
MFCD00082687 CAS Number: 5393-99-7 |
Specification Test Results
Melting point | 92-94°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...