3-Methylbenzamidoxime ≥97%

Supplier: Matrix Scientific

Synonyms: N-Hydroxy-3-methylbenzimidamide

034854-1G 034854-5G
101838-510EA 218.46 USD
101838-510 101838-512
3-Methylbenzamidoxime ≥97%
3-Methylbenzamidoxime

Hazard code(s): Irritant

Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Melting Pt: 87…90 °C
MDL Number: MFCD00833266
CAS Number: 40067-82-1

Order Now

Specification Test Results

Melting point 87-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR