1-(2-Methoxyphenyl)piperazine ≥98%
Supplier: Matrix Scientific
034846-25G
034846-100G
101838-490EA
21.54
USD
101838-490
101838-492
1-(2-Methoxyphenyl)piperazine ≥98%
1-(2-Methoxyphenyl)piperazine
Hazard code(s): Irritant
Formula:
C₁₁H₁₆N₂O Boiling Pt: 110…112 °C (1mm) Melting Pt: 37…40 °C Density: 1.09 g/cm³ (20 °C) |
MDL Number:
MFCD00005958 CAS Number: 35386-24-4 EINECS: 252-537-7 UN: 2923 ADR: 8,III |
Specification Test Results
Boiling point | 110-112°C/1 |
Melting point | 37-40°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...