5-Bromo-2-chloronicotinic acid ≥97%

Supplier: Matrix Scientific

034765-5G 034765-25G 034765-100G
101838-322EA 364.62 USD
101838-322 101838-324 101838-326
5-Bromo-2-chloronicotinic acid ≥97%
5-Bromo-2-chloronicotinic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₃BrClNO₂
Melting Pt: 172…174 °C
MDL Number: MFCD03844847
CAS Number: 29241-65-4

Order Now

Specification Test Results

Melting point 172-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR