4-Bromo-o-toluonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-methylbenzonitrile

034751-5G 034751-25G
101838-290EA 47.69 USD
101838-290 101838-292
4-Bromo-o-toluonitrile ≥98%
4-Bromo-o-toluonitrile

Hazard code(s): Irritant-Harmful

Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 70 °C
MDL Number: MFCD00041461
CAS Number: 67832-11-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 63-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR