2-Phenoxybenzaldehyde

Supplier: Matrix Scientific
Warning

034292-1G 034292-5G
101837-362EA 75.38 USD
101837-362 101837-364
2-Phenoxybenzaldehyde
2-Phenoxybenzaldehyde
Formula: C₁₃H₁₀O₂
MW: 198.22
MDL Number: MFCD00800666
CAS Number: 19434-34-5
EINECS: 243-061-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR