Indole-3-carbaldehyd ≥97%

Supplier: Matrix Scientific

Synonyms: 1H-Indole-3-carbaldehyde, Indole-3-carboxaldehyde

033119-25G 033119-100G
101835-048EA 20 USD
101835-048 101835-050
Indole-3-carbaldehyd ≥97%
Indole-3-carbaldehyd

Hazard code(s): Irritant, Stored under Argon, Store in Freezer

Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 192 - 195 °C
Density: 1.34 g/cm³ (20 °C)
MDL Number: MFCD00005622
CAS Number: 487-89-8
EINECS: 207-665-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR