5-Fluoroindole-2-carboxylic acid

Supplier: Matrix Scientific

032776-1G 032776-5G
101834-412EA 18.46 USD
101834-412 101834-414
5-Fluoroindole-2-carboxylic acid
5-Fluoroindole-2-carboxylic acid
Formula: C₉H₆FNO₂
Melting Pt: 258…259 °C
MDL Number: MFCD00005612
CAS Number: 399-76-8
EINECS: 206-919-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR