5-Aminoquinoline
Supplier: Matrix Scientific
|
Warning
|
032631-5G
032631-25G
101834-136EA
26.15
USD
101834-136
101834-138
5-Aminoquinoline
5-Aminoquinoline
Formula:
C₉H₈N₂ MW: 144.18 g/mol Boiling Pt: 310 °C (1013 hPa) Melting Pt: 107 - 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006797 CAS Number: 611-34-7 EINECS: 210-266-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...