3-Fluoropicolinic acid ≥98%

Supplier: Matrix Scientific

032594-1G 032594-5G
101834-070EA 21.54 USD
101834-070 101834-072
3-Fluoropicolinic acid ≥98%
3-Fluoropicolinic acid

Hazard code(s): Irritant

Formula: C₆H₄FNO₂
Melting Pt: 154 °C (dec.)
MDL Number: MFCD04114108
CAS Number: 152126-31-3

Order Now

Specification Test Results

Melting point 154°C (dec.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR