5-Bromo-2-fluoropyrimidine ≥97%

Supplier: Matrix Scientific

032589-1G 032589-5G
101834-058EA 20 USD
101834-058 101834-060
5-Bromo-2-fluoropyrimidine ≥97%
5-Bromo-2-fluoropyrimidine

Hazard code(s): Irritant

Formula: C₄H₂BrFN₂
MDL Number: MFCD07787364
CAS Number: 62802-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR