5-Chloro-2-fluorobenzotrifluoride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-4-fluoro-3-(trifluoromethyl)benzene, 2-Fluoro-5-chlorobenzotrifluoride, 4-Chloro-1-fluoro-2-(trifluoromethyl)benzene, 5-Chloro-2-fluorobenzotrifluoride

032586-1G 032586-5G
101834-048EA 32.31 USD
101834-048 101834-050
5-Chloro-2-fluorobenzotrifluoride ≥95%
5-Chloro-2-fluorobenzotrifluoride

Hazard code(s): Flammable

Formula: C₇H₃ClF₄
MW: 198.55 g/mol
Boiling Pt: 145 °C
Density: 1.43 g/cm³
MDL Number: MFCD00060655
CAS Number: 89634-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR