3-Fluoropyridine-2-carbonitrile ≥98%

Supplier: Matrix Scientific

032556-1G 032556-5G
101833-988EA 15.38 USD
101833-988 101833-990
3-Fluoropyridine-2-carbonitrile ≥98%
3-Fluoropyridine-2-carbonitrile

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₆H₃FN₂
Melting Pt: 27…30 °C
MDL Number: MFCD06797501
CAS Number: 97509-75-6

Order Now

Specification Test Results

Melting point 27-30°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR