6-Fluoropyridine-2-carbonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Fluoropicolinonitrile, 2-Cyano-6-fluoropyridine

032555-1G 032555-5G
101833-984EA 53.85 USD
101833-984 101833-986
6-Fluoropyridine-2-carbonitrile ≥98%
6-Fluoropyridine-2-carbonitrile

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₆H₃FN₂
MW: 122.1 g/mol
Melting Pt: 32…35 °C
MDL Number: MFCD09263323
CAS Number: 3939-15-9

Order Now

Specification Test Results

Melting point 32-35°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR