5-Bromo-2-ethoxypyridine ≥98%

Supplier: Matrix Scientific

032353-25G 032353-100G
101833-546EA 75.38 USD
101833-546 101833-548
5-Bromo-2-ethoxypyridine ≥98%
5-Bromo-2-ethoxypyridine

Hazard code(s): Irritant

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Melting Pt: 32…36 °C
MDL Number: MFCD00234311
CAS Number: 55849-30-4

Order Now

Specification Test Results

Melting point 32-36°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR