3-Bromo-2-fluorophenol ≥98%
Supplier: Matrix Scientific
Synonyms:
1-Bromo-2-fluoro-3-hydroxybenzene
032349-1G
032349-5G
101833-526EA
41.54
USD
101833-526
101833-528
3-Bromo-2-fluorophenol ≥98%
3-Bromo-2-fluorophenol
Hazard code(s): Irritant, Light Sensitive, Refrigerate
Formula:
C₆H₄BrFO Melting Pt: 52…54 °C |
MDL Number:
MFCD09864673 CAS Number: 156682-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...