2-(Trifluoromethoxy)benzenesulfonamide ≥97%

Supplier: Matrix Scientific

032345-5G 032345-25G
101833-518EA 30.77 USD
101833-518 101833-520
2-(Trifluoromethoxy)benzenesulfonamide ≥97%
2-(Trifluoromethoxy)benzenesulphonamide

Hazard code(s): Irritant

Formula: C₇H₆F₃NO₃S
MW: 241.19 g/mol
Melting Pt: 124…125 °C
MDL Number: MFCD01320751
CAS Number: 37526-59-3
EINECS: 448-450-0

Order Now

Specification Test Results

Melting point 124-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR