5-Chloro-2-hydroxy-3-nitropyridine

Supplier: Matrix Scientific

Synonyms: 5-Chloro-3-nitropyridin-2-ol

032319-1G 032319-5G
101833-460EA 73.85 USD
101833-460 101833-462
5-Chloro-2-hydroxy-3-nitropyridine
5-Chloro-2-hydroxy-3-nitropyridine
Formula: C₅H₃ClN₂O₃
MW: 174.54 g/mol
MDL Number: MFCD00114884
CAS Number: 21427-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR