3-Bromo-5-chlorobenzonitrile ≥97%

Supplier: Matrix Scientific

032287-1G 032287-5G
101833-394EA 26.15 USD
101833-394 101833-396
3-Bromo-5-chlorobenzonitrile ≥97%
3-Bromo-5-chlorobenzonitrile

Hazard code(s): Irritant

Formula: C₇H₃BrClN
Melting Pt: 70…71 °C
MDL Number: MFCD06200840
CAS Number: 304854-55-5

Order Now

Specification Test Results

Melting point 70-71°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR