1H-Pyrazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: 1H-Pyrazol-5-carbaldehyde, 5-Formyl-1H-pyrazole

032273-1G 032273-5G
101833-368EA 58.46 USD
101833-368 101833-370
1H-Pyrazole-5-carbaldehyde ≥95%
1H-Pyrazole-5-carbaldehyde
Formula: C₄H₄N₂O
MW: 96.087 g/mol
Boiling Pt: 300 °C
Melting Pt: 142…144 °C
Density: 1.323 g/cm³
Storage Temperature: Freezer
MDL Number: MFCD06657574
CAS Number: 948552-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR